Mini Nutating Rocker
120 V
Catalog # 470107-056
Supplier: Ward's
Supplier #: H3D-1020
Jump to:
Product Details & Documents
This mini mixer takes up minimal bench space, while providing maximum mixing efficiency.
- Fixed speed
- Small platform
- Flat cord stays out of your way
- 2 year warranty
Capable of supporting loads of up to 4 pounds, this fixed speed orbital mixer is small but powerful. With 3D nutating movement, it will agitate your samples gently, but thoroughly.
Specifications
- Electrical:120 V