Tryptic Soy Broth, Prepared Bottled Media, BD Diagnostics
Catalog # 95062-835
Supplier: BD
CAS Number:
Specifications
- Description:Tryptic Soy Broth, Sterility Bottle
- Size:100 mL
- Cat. No.:95062-835
- Type:Wide Mouth
- Supplier no.:257486
Specifications
About this item
BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples
- Use of highest quality BBL and Difco brand dehydrated culture media for preparation
- Conformation to current USP and EP requirements (see package labeling for details)
- Color-coded caps and/or labeling for most products, providing easy product identification
- Transparent labeling to allow a clear view of media and culture
Wide Mouth bottle holds 100 mL fill volume with lug cap.
They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.