PaintStripe™ Floor Marking Stencil, Brady Worldwide
Supplier: Brady Worldwide
Certificates
About this item
PaintStripe™ Floor Marking Stencils facilitate quick and easy painting of professional quality floor-marking lines on concrete and other industrial surfaces.
- Stencils align to the desired lay-out, masking both sides of the paint line with a single roll of material
- Printed markings on the stencil allow for easy measurement of line distance and rapid alignment of 90° corners
- Can be applied easily by one person working alone
PaintStripe™ Floor Marking Stencils offer a convenient and precise solution wherever painted lines are preferred to create a safe, organized, and productive work environment.