Chart Recorder for Incubators, Thermo Scientific
Catalog # 13306-252
Supplier: Thermo Fisher Scientific
Specifications
- Description:Digital Chart Recorder
- Environmentally Preferable:
- Cat. No.:13306-252
- Supplier no.:3166207
Specifications
About this item
This circular digital chart recorder is designed to use with the automatic water jacketed CO2 incubator, model 815 low-temperature incubators and model 818 diurnal incubators.