Biohazard No Food or Drink to be Stored in this Refrigerator Sign, Brady Worldwide
Supplier: Brady Worldwide
Certificates
About this item
Chemical, Biohazard, and Hazardous Material signs.
- B-401 plastic signs are ideal for use in light-duty industrial, utility, commercial, and institutional environments
- Durable non-reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial, and institutional environments
- Self-Sticking Polyester (B-485) is used to identify the actual or potential presence of a biologically hazardous material
Biohazard indoor/outdoor signs are available in a variety of materials, allowing users to choose the material and size that best fits any application.