PathHunter® eXpress DRD3 U2OS β-Arrestin GPCR Assay
Catalog # 103502-106
Supplier: Eurofins DiscoverX Corporation
CAS Number:
Specifications
- Description:eXpress DRD3 U2OS β-Arrestin GPCR assay
- Size:200 dp (2×96-well)
- Brand Name:PathHunter®
- Cat. No.:103502-106
Specifications
About this item
Access DiscoverX's extensive menu of functional, whole cell GPCR, Kinase, Interleukin, Pathway, NHR, and more assays without investing in a clonal cell line.
- Complete off-the-shelf assay solution
- Pre-validated frozen cells and reagents
- Fast and simple step-by-step protocol
- Compatible with any standard luminometer
Please refer to the product-specific datasheet for the appropriate storage conditions for these products.
Our ultra convenient eXpress™ kits are ready to use and contain all the necessary reagents to perform functional whole cell assays - in just a few hours. Kits include frozen, pre-validated cells, detection reagents, seeding media and plates and do not require expensive instrumentation to run. Minimize assay development time and eliminate lengthy, expensive and time consuming cell culture. Simply thaw, plate and run the assay!