To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Precipitation
- Prep size:Mini
- Refine target molecule:Yeast/Fungal RNA
- Sample type:Yeast/Fungi
- Target molecule:RNA
- Description:MasterPure™ Yeast RNA Purification Kit, Lucigen / Epicentre
- Size:100 RXN
- Description:MasterPure™ Yeast RNA Purification Kit
- Cat. No.:76081-662
Specifications
About this item
Safe, fast purification of high quality RNA from multiple species of yeast
The MasterPure™ Yeast RNA Purification Kit provides all of the reagents needed to purify RNA from yeast cell types (including Candida, Saccharomyces, Schizosaccharomyces) and filamentous fungi. The kit uses a rapid salting-out process to remove contaminating macromolecules, avoiding toxic organic solvents, bead-beating, and spheroplasting.
Safe: Avoids the use of dangerous hot acid phenol
Fast: Quicker protocol that methods using spheroplasting
High Quality RNA: RNA quality exceeds that obtained by bead beating protocols and is ready for use in downstream applications such and RT-PCR and microarray analysis
Excellent Yields: Yields 25-50 µg RNA from 1 ml of mid-log S.cerevisiae
Complete: All the necesseary reagents are supplied with no need to purchase extra equipment