About this item
Quickly purify high yields of high-molecular-weight genomic DNA, total cellular RNA or Total Nucleic Acid (TNA) with one kit
The MasterPure™ Complete DNA and RNA Purification Kit enables rapid, high yield purification of high-molecular-weight genomic DNA, total cellular RNA and Total Nucleic Acid (TNA) from many different sample types.
The kit procedure uses a safe, gentle, and scalable salt-precipitation protocol that eliminates the need for hazardous chemicals and yield-reducing columns. Purification reactions can be scaled up or scaled down depending on the amount of nucleic acid needed and the size of the sample.
Applications include:
Library preparation for next generation sequencing (NGS) of genomic DNA and RNA
DNA methylation studies using Illumina® Infinium® HumanMethylation BeadChips
Genomic DNA and cDNA cloning
qPCR and qRT-PCR
Micrarray analyses (CGH, gene expression profiling, etc)
Fast:Purify Total Nucleic Acid (TNA), DNA or RNA in 30 – 60 minutes
Versatile: Purify TNA, genomic DNA, total RNA, FFPE RNA, or both genomic DNA and total RNA from a sample
Safe: Does not use hazardous phenol, chloroform or guanidine
High Purity: A260/A280 ratios consistently between 1.8 and 2.0
High Yields: Improves yields by avoiding the use of columns which often reduce nucleic acid yields
Total RNA Recovery: Purify both large and small (e.g., miRNA) RNA for RNA-Seq or qRT-PCR
Proven: Hundreds of citations for purification of DNA and RNA from dozens of sample types for use in many applications
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberBiosearch Technologies MasterPure Complete DNA and RNA Purification Kit