Order Entry
Puerto Rico
ContactUsLinkComponent
Nitric acid 5% Blank, Assurance® Grade for ICP, for atomic absorption spectroscopy
Corrosive
Nitric acid 5% Blank, Assurance® Grade for ICP, for atomic absorption spectroscopy
Catalog # SXPLBLK-HNO3
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Corrosive
Nitric acid 5% Blank, Assurance® Grade for ICP, for atomic absorption spectroscopy
Catalog # SXPLBLK-HNO3
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  PLBLK-HNO3
CAS Number:  

Specifications

  • Pack type:
    LDPE bottle
  • Size:
    500 ml
  • Cat. No.:
    SXPLBLK-HNO3

Specifications

About this item

 :
 :63.01 g/mol
UN:3264
ADR:8,III
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349