Hepatitis B Surface Antigen (HBsAg, native or recombinant) ELISA Kit, Alpha Diagnostic
Catalog # MSPP-4110
Supplier: ALPHA DIAGNOSTIC INTERNAT MS
CAS Number:
Specifications
- Primary antibody reactivity:Human
- Sensitivity:0-20 ng/ml
- Cat. No.:MSPP-4110
- Supplier no.:4110
Specifications
About this item
Quantitative ELISA Kit is used for the measurement of recombinant HBsAg in manufacturing process or vaccine formulations.
Caution: For research use only, not for diagnostic or therapeutic use.