Order Entry
Puerto Rico
ContactUsLinkComponent
Nitrite Standard in Water, for Ion Chromatography
Nitrite Standard in Water, for Ion Chromatography
Catalog # ULICC-007
Supplier:  Ultra Scientific
Small Business Enterprise
CAS Number:  
undefined
Nitrite Standard in Water, for Ion Chromatography
Catalog # ULICC-007
Supplier:  Ultra Scientific
Supplier Number:  ICC-007
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 ppm
  • Matrix:
    H₂O
  • Description:
    Nitrite
  • Size:
    125 ml
  • Cat. No.:
    ULICC-007
  • Supplier no.:
    ICC-007

Specifications

About this item