Order Entry
Puerto Rico
ContactUsLinkComponent
Puritan ESK® Environmental Sampling Kit, with Neutralizing Buffer, Sterile
Puritan ESK® Environmental Sampling Kit, with Neutralizing Buffer, Sterile
Catalog # 76045-456
Supplier:  Hardy Diagnostics
undefined
Puritan ESK® Environmental Sampling Kit, with Neutralizing Buffer, Sterile
Catalog # 76045-456
Supplier:  Hardy Diagnostics
Supplier Number:  2583010PDBNB

Specifications

  • Sterility:
    Sterile
  • Description:
    Polyester swabs with 10 ml neutralizing buffer, in tube
  • Size:
    16×100 mm (tube)
  • Cat. No.:
    76045-456
  • Packaging:
    6 boxes of 50 individually wrapped systems (300/case)
  • Fill solution:
    10 ml

Specifications

About this item

Sterile Environment Sampling Kit Pre-Filled with 10 ml Neutralizing Buffer.

  • Easy edge-peel packaging
  • Quick-turn leak-resistant cap
  • High-quality Puritan polyester tipped swab
  • Swab is securely attached to cap
  • Polystyrene handle

Intended for the detection of microorganisms found on dairy and food equipment disinfected with chlorine or quaternary ammonium compounds.

Shelf life - 18 months