Order Entry
Puerto Rico
ContactUsLinkComponent
QuickExtract™ Plant DNA Extraction Solution, PCR-ready DNA from Leaves, Biosearch Technologies
QuickExtract™ Plant DNA Extraction Solution, PCR-ready DNA from Leaves, Biosearch Technologies
Catalog # 76081-772
Supplier:  Lucigen
CAS Number:  
undefined
QuickExtract™ Plant DNA Extraction Solution, PCR-ready DNA from Leaves, Biosearch Technologies
Catalog # 76081-772
Supplier:  Lucigen
Supplier Number:  QEP70750
CAS Number:  

Specifications

  • Method / Format:
    Extraction for direct PCR
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    QuickExtract™ Plant DNA Extraction Solution
  • Size:
    50 ml
  • Description:
    QuickExtract™ Plant DNA Extraction Solution
  • Cat. No.:
    76081-772

Specifications

About this item

Rapid and efficient extraction (no purification) of PCR-ready genomic DNA from plant leaves

  • Fast: PCR-ready DNA in about 8 minutes for most samples
  • Simple: No bead-beating, freezing, or grinding of plant leaf material
  • Increased Yields: No centrifugation or spin columns used which reduce yields
  • Compatible: Extracted DNA is compatible with both real-time and endpoint PCR
  • Safe: Nontoxic reagents used throughout the procedure

The QuickExtract™ Plant DNA Extraction Solution can be used to rapidly and efficiently extract PCR-ready genomic DNA from most plant leaf samples using a simple, one-tube protocol that takes only 8 minutes

The QuickExtract Plant method allows for the inexpensive processing of one to hundreds of samples simultaneously, without grinding the sample, centrifugation, spin columns, or any toxic organic solvent. The procedure is fully compatible with robotic automation, provides a PCR-ready sample, and is reproducible. Simply add the QuickExtract Plant solution to the sample and perform two sequential heating steps. A small aliquot of the sample is then used as a template for PCR or qPCR.

Most leafy plants are suitable for DNA extraction using the QuickExtract Plant Solution, including Arabidopsis, barley, maize, emmer, pepper, rice, spelt, spinach, soybeans, and wheat.