Simple, rapid extraction (no purification) of DNA for PCR-based screening and genotyping type applications
- Fast: 8 minute extraction protocol for most sample types
- Simple: No centrifugation steps or spin columns required
- Automation-friendly: Simple protocol integrated easily into automated workflows
- Safe: Uses only non-toxic reagents
The QuickExtract™ DNA Extraction Solution can be used to rapidly and efficiently extract PCR-ready genomic DNA from almost any sample type using a simple, one-tube protocol that takes only 3-8 minutes, depending on the sample
The QuickExtract method allows for the inexpensive processing of one to hundreds of samples simultaneously, without centrifugation, spin columns or the use of any toxic organic solvent. The method is also compatible with robotic automation.
QuickExtract Solution has been used to extract DNA from samples such as hair follicles, quill-end cells of feathers, tissue-culture cells, buccal cells, zebrafish organs and scales, and mouse tail snips. The extracted DNA is suitable for PCR analyses, such as genomic, transgenic, or viral DNA screening in animals, or for genetic or environmental research and screening in humans and other organisms.
- Method / Format:Extraction for direct PCR
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Rodent tailCultured cellsHuman/animal cells and tissues
- Target molecule:DNA
- Description:QuickExtract™ DNA Extraction Solution