Order Entry
Puerto Rico
ContactUsLinkComponent
Rec J Exonuclease
Rec J Exonuclease
Catalog # 76081-688
Supplier:  Lucigen
CAS Number:  
undefined
Rec J Exonuclease
Catalog # 76081-688
Supplier:  Lucigen
Supplier Number:  RJ411250
CAS Number:  

Specifications

  • Size:
    250 units
  • Enzyme Name:
    Rec J Exonuclease
  • Concentration:
    10 units/μl
  • Cat. No.:
    76081-688

Specifications

About this item

Magnesium-dependent digestion of single-stranded DNA in a 5´ to 3´ direction

  • Specific: Digests ssDNA in the 5' to 3 direction
  • Heat Inactivated: Heat at 65°C for 20 minutes to kill enzyme activity
  • Flexible: Use to remove primers from completed PCR or to degrade single-stranded linear DNA in dsDNA and plasmid preps

Rec J Exonuclease, derived from E

Applications include:
Removal of primers from completed PCR
Degradation of single-stranded linear DNA in dsDNA and plasmid preps

coli, catalyzes degradation of ssDNA in a 5´ to 3´ direction. Its activity is dependent on Mg2+. Rec J Exonuclease can be heat-inactivated by incubation at 65°C for 20 minutes.

Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and a 10X reaction buffer.