Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
DNase

DNase

5000 units
Catalog # 76081-624
Supplier:   Lucigen
Supplier #:  DB0715K
DNase
Catalog # 76081-624
Supplier:  Lucigen
CAS Number:  
DNase
DNase

5000 units

CAS Number:  
Price
$708.48
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Effectively digest large or small dsDNA and ssDNA into mononucleotides

  • Digest unwanted dsDNA and ssDNA molecules including small ssDNA such as random hexamers
  • Use in sensitive applications requiring reverse transcription where any contaminating DNA is unwanted

Baseline-ZERO™ DNase digests dsDNA and ssDNA into mononucleotides more effectively than the commonly used bovine pancreatic DNase I

Applications include:
Removal of genomic DNA from small-sample total RNA preparations for expression analysis
Removal of small DNA oligonucleotides (e.g., random primers)

Even the small DNA oligonucleotides that remain after treatment with bovine pancreatic DNase I are undetectable by gel electrophoresis following treatment with Baseline-ZERO DNase. Removal of DNA from RNA preparations is particularly beneficial when RNA in a sample is amplified using a method that involves reverse transcription using random primers, since any contaminating DNA would also be a template for random-primed cDNA synthesis.

Certifications: For research use only. Not for human or diagnostic use.

Ordering information: Contents include enzyme and a 10X reaction buffer. Units reported are MBU or Molecular Biology Units. See the product user manual for the definition of MBU.

Documents

Specifications

Specifications

  • Size:
    5000 units
  • Cat. No.:
    76081-624
  • Concentration:
    1 units/μl
  • Enzyme Name:
    DNase

Specifications