DNase I
Degrade dsDNA and ssDNA without damaging important RNA in your sample or reaction
- Digest away unwanted DNA without digesting RNA
- Use in a variety of applications such as in vitro transcription and RT-PCR
RNase-Free DNase I (bovine pancreas) is an endonuclease useful in removing DNA that might interfere with the characterization, manipulation, or use of RNA, or for any application requiring highly purified DNase I, such as nick translation
Applications include:
Elimination of template DNA following in vitro synthesis of RNA with T7, SP6, or T3 RNA polymerase
Labeling of DNA by nick translation, in combination with Klenow or other DNA polymerases
Treatment of RNA prior to RT-PCR
Characterization of DNA-protein interactions by DNase I footprinting
It efficiently hydrolyzes dsDNA and ssDNA into a mixture of short oligonucleotides and mononucleotides.
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and a 10X reaction buffer. Units reported are MBU or Molecular Biology Units. See the product user manual for the definition of MBU.
Degrade dsDNA and ssDNA without damaging important RNA in your sample or reaction
- Digest away unwanted DNA without digesting RNA
- Use in a variety of applications such as in vitro transcription and RT-PCR
RNase-Free DNase I (bovine pancreas) is an endonuclease useful in removing DNA that might interfere with the characterization, manipulation, or use of RNA, or for any application requiring highly purified DNase I, such as nick translation
Applications include:
Elimination of template DNA following in vitro synthesis of RNA with T7, SP6, or T3 RNA polymerase
Labeling of DNA by nick translation, in combination with Klenow or other DNA polymerases
Treatment of RNA prior to RT-PCR
Characterization of DNA-protein interactions by DNase I footprinting
It efficiently hydrolyzes dsDNA and ssDNA into a mixture of short oligonucleotides and mononucleotides.
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and a 10X reaction buffer. Units reported are MBU or Molecular Biology Units. See the product user manual for the definition of MBU.
Documents
- Catalog No:
- 76081-620
- 76081-622
- Enzyme Name:
- DNase I
- DNase I
- Concentration:
- 1 units/μl
- 1 units/μl
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



