Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Rat Recombinant IL-2 (from <i>E. coli</i>)

Rat Recombinant IL-2 (from E. coli)

Rat Recombinant IL-2 (from <i>E. coli</i>)
Certificates
Rat Recombinant IL-2 (from <i>E. coli</i>)

4 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes.  IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.

  • High Quality
  • Low Endotoxin
  • Affordable

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 75845-232
  • 75845-234
  • 75845-300
  • 75845-302
  • 75845-304
  • Protein/Peptide Name:
  • IL-2
  • IL-2
  • IL-2
  • IL-2
  • IL-2
  • Protein Synonyms:
  • Aldesleukin|TCGF|T cell growth factor
  • Aldesleukin|TCGF|T cell growth factor
  • Aldesleukin|TCGF|T cell growth factor
  • Aldesleukin|TCGF|T cell growth factor
  • Aldesleukin|TCGF|T cell growth factor
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Rat
  • Rat
  • Rat
  • Rat
  • Rat
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P17108
  • P17108
  • P17108
  • P17108
  • P17108
  • Sequence:
  • MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
  • MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
  • MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
  • MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
  • MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-Free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-Free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin Level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin Content:
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
  • Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
  • Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
  • Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
  • Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
  • Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Formulation:
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • Molecular Weight:
  • 15.6 kDa
  • 15.6 kDa
  • 15.6 kDa
  • 15.6 kDa
  • 15.6 kDa
  • Reconstitution Instructions:
  • Sterile 10 mM HCl at 0.1 mg/ml
  • Sterile 10 mM HCl at 0.1 mg/ml
  • Sterile 10 mM HCl at 0.1 mg/ml
  • Sterile 10 mM HCl at 0.1 mg/ml
  • Sterile 10 mM HCl at 0.1 mg/ml
  • Storage Conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping Temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • Tested Applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Origin
    Availability
    Price
  • Vial
    2 μg
    US
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    75845-232
    Protein/Peptide Name
    IL-2
    Protein Synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P17108
    Sequence
    MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    Form
    Lyophilized
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    15.6 kDa
    Reconstitution Instructions
    Sterile 10 mM HCl at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    20 µg
    Log in for availability
    Log in for availability
    Each (20µG)
    Each (20µG) - $218.67
    $218.67
    Each (20µG)
       $218.67
    Minimum order quantity is 1

    Supplier Number: 300-38-20UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    75845-234
    Protein/Peptide Name
    IL-2
    Protein Synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P17108
    Sequence
    MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Form
    Lyophilized
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    15.6 kDa
    Reconstitution Instructions
    Sterile 10 mM HCl at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    100 µg
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $700.43
    $700.43
    Each (100µG)
       $700.43
    Minimum order quantity is 1

    Supplier Number: 300-38-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    75845-300
    Protein/Peptide Name
    IL-2
    Protein Synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P17108
    Sequence
    MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Form
    Lyophilized
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    15.6 kDa
    Reconstitution Instructions
    Sterile 10 mM HCl at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    500 µg
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $2,446.90
    $2,446.90
    Each (500µG)
       $2,446.90
    Minimum order quantity is 1

    Supplier Number: 300-38-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    75845-302
    Protein/Peptide Name
    IL-2
    Protein Synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P17108
    Sequence
    MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Form
    Lyophilized
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    15.6 kDa
    Reconstitution Instructions
    Sterile 10 mM HCl at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $3,935.13
    $3,935.13
    Each (1mg)
       $3,935.13
    Minimum order quantity is 1

    Supplier Number: 300-38-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    75845-304
    Protein/Peptide Name
    IL-2
    Protein Synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P17108
    Sequence
    MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Form
    Lyophilized
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    15.6 kDa
    Reconstitution Instructions
    Sterile 10 mM HCl at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.