Rat Recombinant M-CSF (from E. coli)
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types. M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Rat
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity Assay: NFS-60 Proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1.0×10⁵ units/mg
- Gene ID:Q8JZQ0
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:CSF-1|mgI-IM
- Protein/Peptide Name:M-CSF
- Purity:≥95%
- Molecular Weight:18.1/36.2 kDa
- Sequence:MEVSEHCSHM IGNGHLQILQ QLIDSQMETA CLIEYKFVDQ EQLDDPVCYL KKAFVLVQVI IEETMRFKDN TPNANATERL QELSMKLNSC FIKDYKEQNE ACVQTYKESP LRLLEKIKNF FNETKNFLEK DWNIFSKNCN DSLAKCSSRD VVTKP
- Endotoxin Level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay