Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-17A (from E. coli)
Human Recombinant IL-17A (from E. coli)
Catalog # 10822-158
CAS Number:  
undefined
Human Recombinant IL-17A (from E. coli)
Catalog # 10822-158
Supplier Number:  100-87-500UG
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    500 µg
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Production of IL-6 from mouse 3T3 cells, ED50 ≤10 ng/ml, bioactivity ≥1.0×10⁵ units/mg
  • Form:
    Lyophilized
  • Gene ID:
    Q16552
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    IL-17|CTLA-8
  • UniProtKB:
    Q16552
  • Protein/Peptide Name:
    IL-17A
  • Purity:
    ≥95%
  • Molecular Weight:
    15.7/31.3 kDa
  • Sequence:
    MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVA
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay
  • Cat. No.:
    10822-158
  • Supplier no.:
    100-87-500UG

Specifications

About this item

Interleukin 21 (IL-21) is a member of the common-gamma chain family of cytokines that are involved in immunoregulation. IL-21 is normally expressed by activated CD4+ T cells and is aberrantly expressed in Hodgkin lymphoma cells. The IL-21 receptor (IL-21R) activates the JAK/STAT signaling pathway and is expressed on T, B, and natural killer (NK) cells.

  • High quality
  • Low endotoxin
  • Affordable

Within the B cell lineage, IL-21 is a switch factor regulating IgG1 and IgG3 antibody production. IL-21 also cooperates with IL-4 for the production of multiple antibody classes in B cells. IL-21 has pleiotropic effects on the proliferation, differentiation, and effector functions of B, T, NK, and dendritic cells.

Caution: For research use only.