Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-11 (Animal free) (from E. coli)
Human Recombinant IL-11 (Animal free) (from E. coli)
Certificates
undefined
Human Recombinant IL-11 (Animal free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: TF-1 Cell Proliferation, ED50 ≤10 ng/ml, bioactivity ≥1.0×10⁵ units/mg
  • Form:
    Lyophilized
  • Gene ID:
    P20809
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    oprelvekin|AGIF (Adipogenesis Inhibitory Factor)
  • UniProtKB:
    P20809
  • Protein/Peptide Name:
    IL-11 (AF)
  • Purity:
    ≥95%
  • Molecular Weight:
    19.3 kDa
  • Sequence:
    MPGPPPGPPR VSPDPRAELD STVLLTRSLL ADTRQLAAQL RDKFPADGDH NLDSLPTLAM SAGALGALQL PGVLTRLRAD LLSYLRHVQW LRRAGGSSLK TLEPELGTLQ ARLDRLLRRL QLLMSRLALP QPPPDPPAPP LAPPSSAWGG IRAAHAILGG LHLTLDWAVR GLLLLKTRL
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications

Related Information