Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-5 (from E. coli)
Human Recombinant IL-5 (from E. coli)
Certificates
undefined
Human Recombinant IL-5 (from E. coli)
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤0.25 ng/ml, Bioactivity ≥4.0×10⁶ units/mg
  • Form:
    Lyophilized
  • Gene ID:
    P05113
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    Eosinophil differentiation factor|TRF|B-cell differentiation factor I
  • Protein/Peptide Name:
    IL-5
  • Purity:
    ≥95%
  • Molecular Weight:
    13.3/26.6 kDa
  • Sequence:
    MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium glycinate, pH 8.5
  • Nuclease-free:
    N
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay
  • Grade:
    RUO

Specifications

Related Information