Human Recombinant GACRP-30 (from E. coli)
Supplier: Fujifilm Irvine Scientific
New Product
Certificates
About this item
The globular subunit of adipocyte complement-related protein of 30 kDa (ACRP-30) is a naturally occurring cleavage product of adiponectin, a protein made exclusively by adipocytes. ACRP-30 is an abundant serum protein and plays an important role in hyperglycemia, insulin resistance, and fatty acid oxidation. ACRP-30 signals through adiponectin receptor 1 (AdipoR1) and adiponectin receptor 2 (AdipoR2).
- High quality
- Low endotoxin
- Affordable
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity Assay: Inhibition of M1 cell proliferation, ED₅₀ ≤2000 ng/ml, Bioactivity ≥500 units/mg
- Gene ID:Q15848
- Reconstitution Instructions:Sterile 10 mM sodium phosphate, 0.5 mM DTT, pH 7.5 at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:adipolean|Adiponectin|apm-1
- Protein/Peptide Name:GACRP-30 (AF)
- Purity:≥90%
- Molecular Weight:16.7 kDa
- Sequence:MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN
- Endotoxin Level:Low
- Formulation:10 mM Sodium phosphate, 0.5 mM DTT, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Frequently Bought Together