Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant IGF-II (Animal free) (from E. coli)
 
undefined
Human Recombinant IGF-II (Animal free) (from E. coli)
 

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7, ED50 ≤6 ng/ml, bioactivity ≥1.7×10⁵ units/mg
  • Form:
    Lyophilized
  • Gene ID:
    P01344
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Somatomedin A
  • UniProtKB:
    P01344
  • Protein/Peptide Name:
    IGF-II (AF)
  • Purity:
    ≥95%
  • Molecular Weight:
    7.5 kDa
  • Sequence:
    AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

 

Frequently Bought Together