Order Entry
Puerto Rico
ContactUsLinkComponent
5'-Deoxyadenosine, [5'-3H] ≥97% (by HPLC), MORpure™
5'-Deoxyadenosine, [5'-3H] ≥97% (by HPLC), MORpure™
Catalog # 101973-494
CAS Number:  
undefined
5'-Deoxyadenosine, [5'-3H] ≥97% (by HPLC), MORpure™
Catalog # 101973-494
Supplier Number:  MT718-250UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    250 µCi
  • Cat. No.:
    101973-494
  • Supplier no.:
    MT718-250UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:2086327-58-2