Order Entry
Puerto Rico
ContactUsLinkComponent
Sucrose, [3H] ≥97% (by HPLC), MORpure™
Irritant
Sucrose, [3H] ≥97% (by HPLC), MORpure™
Catalog # 101972-866
CAS Number:  
undefined
Irritant
Sucrose, [3H] ≥97% (by HPLC), MORpure™
Catalog # 101972-866
Supplier Number:  MT1956-50UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    50 µCi
  • Cat. No.:
    101972-866
  • Supplier no.:
    MT1956-50UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:112642-98-5