Order Entry
Puerto Rico
ContactUsLinkComponent
2'-Deoxy-2'-fluoroarabinofuranosylcytosine, [3H] ≥97% (by HPLC), MORpure™
Irritant
2'-Deoxy-2'-fluoroarabinofuranosylcytosine, [3H] ≥97% (by HPLC), MORpure™
Catalog # 101972-654
CAS Number:  
undefined
Irritant
2'-Deoxy-2'-fluoroarabinofuranosylcytosine, [3H] ≥97% (by HPLC), MORpure™
Catalog # 101972-654
Supplier Number:  MT1858-250UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    250 µCi
  • Cat. No.:
    101972-654
  • Supplier no.:
    MT1858-250UCI

Specifications

About this item

 :
ADR:,
 :245.21 g/mol
UN:2910
 :GHS07
CAS Number:56632-83-8
Formula:C₉H₁₂FN₃O₄
MDL Number:MFCD07374399