
Estriol, [2,4-3H] ≥97% (by HPLC), MORpure™
Catalog # 101972-618
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:NextGen V-Vial
- Size:250 µCi
- Cat. No.:101972-618
- Supplier no.:MT1785-250UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:19590-79-5