Order Entry
Puerto Rico
ContactUsLinkComponent
5-Hydroxymethyl-2'-deoxyuridine, [3H] ≥97% (by HPLC), MORpure™
Irritant
5-Hydroxymethyl-2'-deoxyuridine, [3H] ≥97% (by HPLC), MORpure™
Catalog # 101972-606
CAS Number:  
undefined
Irritant
5-Hydroxymethyl-2'-deoxyuridine, [3H] ≥97% (by HPLC), MORpure™
Catalog # 101972-606
Supplier Number:  MT1781-250UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    250 µCi
  • Cat. No.:
    101972-606
  • Supplier no.:
    MT1781-250UCI

Specifications

About this item

 :
 :258.228 g/mol
UN:2910
ADR:,
 :GHS07
CAS Number:5116-24-5
Formula:C₁₀H₁₄N₂O₆
MDL Number:MFCD00047489