Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Deazaadenosine, [3H]
3-Deazaadenosine, [3H] ≥97% (by HPLC), MORpure™
3-Deazaadenosine, [3H] ≥97% (by HPLC), MORpure™
Irritant
3-Deazaadenosine, [3H] ≥97% (by HPLC), MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
Irritant
3-Deazaadenosine, [3H] ≥97% (by HPLC), MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
339155-12-3
:
:
266.25 g/mol
UN:
2910
ADR:
,
:
GHS07