Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Caffeine, [8-14C]
Caffeine, [8-14C] ≥98% (by HPLC), MORpure™
Caffeine, [8-14C] ≥98% (by HPLC), MORpure™
Irritant
Caffeine, [8-14C] ≥98% (by HPLC), MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
Irritant
Caffeine, [8-14C] ≥98% (by HPLC), MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
61603-17-6
:
UN:
2910
ADR:
,
:
GHS07