
Molinate, [hexahydroazepine-2-14C] ≥98% (by HPLC), MORpure™
Catalog # 101971-500
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:Ampoule, sealed
- Size:10 µCi
- Cat. No.:101971-500
- Supplier no.:MC2376-10UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:13513-98-9