
Z-D,L-Serine, [3-14C] ≥97% (by HPLC)
Catalog # 101971-456
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:NextGen V-Vial
- Size:50 µCi
- Cat. No.:101971-456
- Supplier no.:MC2344-50UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:2088204-21-9