
Docetaxel, [tert-butyl-14C] ≥98% (by HPLC), MORpure™
Catalog # 101971-422
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:NextGen V-Vial
- Size:10 µCi
- Cat. No.:101971-422
- Supplier no.:MC2333-10UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:2088204-07-1