
Tris(chloroisopropyl)phosphate, [propyl-1,2-14C] ≥98% (by HPLC), MORpure™
Catalog # 101971-348
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:Ampoule, sealed
- Size:50 µCi
- Cat. No.:101971-348
- Supplier no.:MC2292-50UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:2088206-85-1