$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
D-Glucose, [1-14C]
D-Glucose, [1-14C] ≥98% (by HPLC), MORpure™
D-Glucose, [1-14C] ≥98% (by HPLC), MORpure™
Irritant
D-Glucose, [1-14C] ≥98% (by HPLC), MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
Irritant
D-Glucose, [1-14C] ≥98% (by HPLC), MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
910906-34-2
:
UN:
2910
ADR:
,
:
GHS07