Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pentane-2,4-dione, [2,4-14C]
Pentane-2,4-dione, [2,4-14C] ≥98% (by HPLC), MORpure™
Pentane-2,4-dione, [2,4-14C] ≥98% (by HPLC), MORpure™
Pentane-2,4-dione, [2,4-14C] ≥98% (by HPLC), MORpure™
101971-332
:
MORAVEK BIOCHEMICALS MS
:
Pentane-2,4-dione, [2,4-14C] ≥98% (by HPLC), MORpure™
101971-332
:
MORAVEK BIOCHEMICALS MS
:
MC2288-250UCI
:
Pack type:
Ampoule, sealed
Size:
250 µCi
Cat. No.:
101971-332
Supplier no.:
MC2288-250UCI
:
:
2910
:
,
:
GHS07
:
70838-66-3