Order Entry
Puerto Rico
ContactUsLinkComponent
(S)-HPMPA, [adenine-8-14C] ≥98% (by HPLC), MORpure™
Irritant
(S)-HPMPA, [adenine-8-14C] ≥98% (by HPLC), MORpure™
Catalog # 101971-300
CAS Number:  
undefined
Irritant
(S)-HPMPA, [adenine-8-14C] ≥98% (by HPLC), MORpure™
Catalog # 101971-300
Supplier Number:  MC2273-50UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    50 µCi
  • Cat. No.:
    101971-300
  • Supplier no.:
    MC2273-50UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:2088573-53-7