
3-Chlorobenzoic acid, [ring-14C(U)] ≥98% (by HPLC), MORpure™
Catalog # 101971-222
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:NextGen V-Vial
- Size:250 µCi
- Cat. No.:101971-222
- Supplier no.:MC2225-250UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:2088575-34-0