Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Octanoyl coenzyme A
Octanoyl coenzyme A
Octanoyl coenzyme A
Irritant
Octanoyl coenzyme A
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
Irritant
Octanoyl coenzyme A
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
1264-52-4
:
Formula:
C₂₉H₅₀N₇O₁₇P₃S
MDL Number:
MFCD02259055
:
893.730 g/mol
UN:
2910
ADR:
,
:
GHS07