
4-Chlorophenol,[14C(U)] ≥97% (by HPLC)
Catalog # 101971-032
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:NextGen V-Vial
- Size:50 µCi
- Cat. No.:101971-032
- Supplier no.:MC2123-50UCI
Specifications
About this item
:
UN:2910
ADR:,
:GHS07
CAS Number:181125-81-5