$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Sodium hydrogen carbonate-[14C]
Sodium hydrogen carbonate-[14C]
Sodium hydrogen carbonate-[14C]
:
MORAVEK BIOCHEMICALS MS
Sodium hydrogen carbonate-[14C]
:
MORAVEK BIOCHEMICALS MS
:
7279-86-9
:
:
NaHCO₃
:
86
:
2910
:
,
:
GHS07