$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
L-Leucine, [14C(U)]
L-Leucine, [14C(U)] ≥98% (by HPLC), MORpure™
L-Leucine, [14C(U)] ≥98% (by HPLC), MORpure™
Irritant
L-Leucine, [14C(U)] ≥98% (by HPLC), MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
Irritant
L-Leucine, [14C(U)] ≥98% (by HPLC), MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
5155-21-5
:
UN:
2910
ADR:
,
:
GHS07