$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
D-Galactose, [1-14C]
D-Galactose, [1-14C] ≥97% (by HPLC)
D-Galactose, [1-14C] ≥97% (by HPLC)
D-Galactose, [1-14C] ≥97% (by HPLC)
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
D-Galactose, [1-14C] ≥97% (by HPLC)
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
5160-32-7
:
UN:
2910
ADR:
,
:
GHS07