Order Entry
Puerto Rico
ContactUsLinkComponent
Pirfenidone, [14C] ≥98% (by HPLC), MORpure™
Irritant
Pirfenidone, [14C] ≥98% (by HPLC), MORpure™
Catalog # 101970-560
CAS Number:  
undefined
Irritant
Pirfenidone, [14C] ≥98% (by HPLC), MORpure™
Catalog # 101970-560
Supplier Number:  MC1435-50UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    50 µCi
  • Cat. No.:
    101970-560
  • Supplier no.:
    MC1435-50UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:2088140-75-2