Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ciprofloxacin, [2-14C]
Ciprofloxacin, [2-14C] ≥97% (by HPLC)
Ciprofloxacin, [2-14C] ≥97% (by HPLC)
Irritant
Ciprofloxacin, [2-14C] ≥97% (by HPLC)
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
Irritant
Ciprofloxacin, [2-14C] ≥97% (by HPLC)
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
333382-19-7
:
UN:
2910
ADR:
,
:
GHS07