Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Trifluoromethyluracil, [2-14C]
5-Trifluoromethyluracil, [2-14C] ≥98% (by HPLC), MORpure™
5-Trifluoromethyluracil, [2-14C] ≥98% (by HPLC), MORpure™
5-Trifluoromethyluracil, [2-14C] ≥98% (by HPLC), MORpure™
101970-408
:
MORAVEK BIOCHEMICALS MS
:
5-Trifluoromethyluracil, [2-14C] ≥98% (by HPLC), MORpure™
101970-408
:
MORAVEK BIOCHEMICALS MS
:
MC1382-50UCI
:
Pack type:
NextGen V-Vial
Size:
50 µCi
Cat. No.:
101970-408
Supplier no.:
MC1382-50UCI
:
:
2910
:
,
:
GHS07
:
10028-67-8