Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Hexadecanol [1-14C]
1-Hexadecanol [1-14C] ≥98% (by HPLC), MORpure™
1-Hexadecanol [1-14C] ≥98% (by HPLC), MORpure™
:
MORAVEK BIOCHEMICALS MS
1-Hexadecanol [1-14C] ≥98% (by HPLC), MORpure™
:
MORAVEK BIOCHEMICALS MS
:
3877-54-1
:
:
2910
:
,
:
GHS07