$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Potassium cyanate, [14C]
Potassium cyanate, [14C] ≥96% (by HPLC)
Potassium cyanate, [14C] ≥96% (by HPLC)
Irritant
Potassium cyanate, [14C] ≥96% (by HPLC)
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
Irritant
Potassium cyanate, [14C] ≥96% (by HPLC)
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
67877-97-8
:
UN:
2910
ADR:
,
:
GHS07