Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2',3'-Didehydro-3'-deoxythymidine
2',3'-Didehydro-3'-deoxythymidine
2',3'-Didehydro-3'-deoxythymidine
2',3'-Didehydro-3'-deoxythymidine
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
2',3'-Didehydro-3'-deoxythymidine
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
3056-17-5
:
Formula:
C₁₀H₁₂N₂O₄
MDL Number:
MFCD00132921
:
224.22 g/mol